Learn More
Invitrogen™ Phospho-Btk (Tyr223) Recombinant Rabbit Monoclonal Antibody (BtkY223-B4), FITC
Rabbit Recombinant Monoclonal Antibody
$245.00 - $581.50
Specifications
Antigen | Phospho-Btk (Tyr223) |
---|---|
Clone | BtkY223-B4 |
Content And Storage | 4°C, store in dark |
Applications | Flow Cytometry |
Classification | Recombinant Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA528076
|
Invitrogen™
MA528076 |
100 Tests |
Each for $581.50
|
|
|||||
PIMA536925
|
Invitrogen™
MA536925 |
10 Tests |
Each for $245.00
|
|
|||||
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Bruton tyrosine kinase (BTK) is a cytoplasmic tyrosine kinase belonging to the SRC-related TEC subfamily of tyrosine kinases. Mutations in the BTK gene have been linked to severe developmental blocks in human B-cell ontogeny and immunodeficiency disorders. It has recently been shown to interact with members of the toll-like receptor (TLR) family such as TLR4, 6, 8, and 9. The TLRs are critical molecules in both the innate and adaptive immunity and can recognize diverse microbial pathogens. BTK has also been shown to interact with key proteins involved in TLR4 signal transduction such as MyD88, TIRAP, and IRAK, but not TRAF-6, suggesting that BTK is involved in lipopolysaccharide signal transduction.Specifications
Phospho-Btk (Tyr223) | |
4°C, store in dark | |
Recombinant Monoclonal | |
Liquid | |
IgG κ | |
Human, Mouse | |
BTK | |
agammaglobulinaemia tyrosine kinase; agammaglobulinemia tyrosine kinase; AGMX1; AI528679; AT; ATK; B-cell progenitor kinase; BPK; Bruton agammaglobulinemia tyrosine kinase; Bruton tyrosine kinase; Bruton's tyrosine kinase; BTK; dominant-negative kinase-deficient Brutons tyrosine kinase; IMD1; Kinase EMB; MGC126261; MGC126262; OTTHUMP00000023677; PSCTK1; RP1-164F3.2; truncated Bruton agammaglobulinemia tyrosine kinase; tyrosine-protein kinase BTK; Tyrosine-protein kinase BTK (Brutons tyrosine kinase) (Agammaglobulinaemia tyrosine kinase) (ATK) (B cell progenitor kinase) (BPK) (Kinase EMB); tyrosine-protein kinase BTK isoform (lacking exon 13 to 17); tyrosine-protein kinase BTK isoform (lacking exon 14); xid; XLA; X-linked immune deficiency | |
BTK | |
Primary | |
Protein A/G |
BtkY223-B4 | |
Flow Cytometry | |
FITC | |
Rabbit | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P35991, Q06187 | |
12229, 695 | |
A synthetic phospho-peptide corresponding to residues surrounding Tyr223 of human phospho Btk. | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.