Learn More
Invitrogen™ Phospho-c-Abl (Thr735) Polyclonal Antibody

Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with other related proteins.
Specifications
Specifications
| Antigen | Phospho-c-Abl (Thr735) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | 0.01M HEPES with 100 μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 |
| Gene | ABL1 |
| Gene Accession No. | P00519 |
| Gene Alias | Abelson murine leukemia oncogene; Abelson murine leukemia viral (v-abl) oncogene homolog 1; abelson murine leukemia viral oncogene homolog 1; Abelson tyrosine-protein kinase 1; Abl; Abl 1 antib; ABL proto-oncogene 1, non-receptor tyrosine kinase; Abl1; ABL2; ABLL; AI325092; ARG; B30; bcr/abl; bcr/c-abl oncogene protein; c-ABL; c-abl oncogene 1, non-receptor tyrosine kinase; c-abl oncogene 1, receptor tyrosine kinase; c-ABL1; E430008G22Rik; JTK7; p150; Proto-oncogene c-Abl; proto-oncogene tyrosine-protein kinase ABL1; RP11-83J21.1; Tyrosine kinase ARG; tyrosine-protein kinase ABL1; v-abl; v-abl Abelson murine leukemia oncogene 1; v-abl Abelson murine leukemia viral oncogene 1; v-abl Abelson murine leukemia viral oncogene homolog 1 |
| Gene Symbols | ABL1 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.