Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-c-Abl (Tyr89, Tyr70) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564565
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564565 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564565 Supplier Invitrogen™ Supplier No. PA564565
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-c-Abl (Tyr89, Tyr70) Polyclonal Antibody detects endogenous levels of c-Abl only when phosphorylated at Tyr89, Tyr70.

The ABL1 proto-oncogene encodes a cytoplasmic and nuclear protein tyrosine kinase that has been implicated in processes of cell differentiation, cell division, cell adhesion, and stress response. Activity of c-Abl protein is negatively regulated by its SH3 domain, and deletion of the SH3 domain turns c-Abl into an oncogene. The DNA-binding activity of the ubiquitously expressed ABL1 tyrosine kinase is regulated by CDC2-mediated phosphorylation, suggesting a cell cycle function for c-Abl. In chronic myelogenous leukemia and a subset of acute lymphoblastic leukemias, the c-Abl proto oncogene undergoes a (9;22) chromosomal translocation producing a novel rearranged chromosome (the Philadelphia chromosome). As the result of the fusion of c-Abl sequences from chromosome 9 to the Bcr gene on chromosome 22. The c-Abl oncogene was initially identified as the viral transforming gene of Abelson murine leukemia virus (A-MuLV). c-ABL potentially regulates DNA repair by activating the proapoptotic pathway when the DNA damage is too severe to be repaired.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-c-Abl (Tyr89, Tyr70)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ABL1
Gene Accession No. P00519, P00520
Gene Alias Abelson murine leukemia oncogene; Abelson murine leukemia viral (v-abl) oncogene homolog 1; abelson murine leukemia viral oncogene homolog 1; Abelson tyrosine-protein kinase 1; Abl; Abl 1 antib; ABL proto-oncogene 1, non-receptor tyrosine kinase; Abl1; ABL2; ABLL; AI325092; ARG; B30; bcr/abl; bcr/c-abl oncogene protein; c-ABL; c-abl oncogene 1, non-receptor tyrosine kinase; c-abl oncogene 1, receptor tyrosine kinase; c-ABL1; E430008G22Rik; JTK7; p150; Proto-oncogene c-Abl; proto-oncogene tyrosine-protein kinase ABL1; RP11-83J21.1; Tyrosine kinase ARG; tyrosine-protein kinase ABL1; v-abl; v-abl Abelson murine leukemia oncogene 1; v-abl Abelson murine leukemia viral oncogene 1; v-abl Abelson murine leukemia viral oncogene homolog 1
Gene Symbols ABL1
Host Species Rabbit
Immunogen A synthesized peptide derived from human ABL1/2 (Phospho-Tyr89/Tyr70).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11350, 25, 311860
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.