Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-c-Fos (Ser32) Recombinant Rabbit Monoclonal Antibody (cFOSS32-BA9), APC, Invitrogen™
GREENER_CHOICE

Catalog No. PIMA537176
Change view
Click to view available options
Quantity:
10 Tests
100 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537176 100 Tests
PIMA537424 10 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA537176 Supplier Invitrogen™ Supplier No. MA537176
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

FBJ murine osteosarcoma viral oncogene homolog is commonly known as Cellular protooncogene Fos (c-Fos). It belongs to the fos transcription factor family and is encoded by the C-FOS gene located on chromosome 14. c-Fos heterodimerizes with c-Jun to form the Activator Protein -1 (AP-1) transcription factor which regulates transcription of several genes that play an important role in cellular signal transduction, cell proliferation and differentiation. c-Fos has been shown to be phosphorylated by various stimuli including growth factors and insulin at atleast seven different sites. The best known upstream kinase for phosphorylation at Threonine 232 is ERK MAPK which regulates localization of c-Fos to the nucleus and is thus important for c-Fos induced transcriptional activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-c-Fos (Ser32)
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone cFOSS32-BA9
Conjugate APC
Formulation PBS with 0.20% BSA and 0.09% sodium azide
Gene FOS
Gene Accession No. P01100, P01101, P12841
Gene Alias activator protein 1; AP-1; cellular oncogene c-fos; cellular oncogene fos; cFos; C-FOS; c-fos oncogene; c-fos protooncogene protein; D12Rfj1; FBJ murine osteosarcoma viral (v-fos) oncogene homolog (oncogene FOS); FBJ murine osteosarcoma viral oncogene homolog; FBJ murine osteosarcoma viral oncogene homolog B; FBJ osteosarcoma oncogene; FBJ osteosarcoma oncogene B; FBJ osteosarcoma viral oncogene isoform deltaFosb-2; FOS; Fos proto-oncogene, AP-1 trancription factor subunit; Fos proto-oncogene, AP-1 transcription factor subunit; Fosb; FosB proto-oncogene, AP-1 transcription factor subunit; fra-2; G0/G1 switch regulatory protein 7; G0S7; I79_009428; p55; protein fosB; proto-oncogene cFOS; proto-oncogene c-Fos; proto-oncogene protein c-fos; v-fos FBJ murine osteosarcoma viral oncogene homolog
Gene Symbols FOS
Host Species Rabbit
Immunogen A synthetic phospho-peptide corresponding to residues surrounding Ser32 of human phospho c-Fos.
Purification Method Protein A/G
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14281, 2353, 314322
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.