Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-c-Fos (Ser374) Polyclonal Antibody, Cyanine3, Bioss
SDP

Catalog No. 502281342
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-1342 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-1342 Supplier Bioss Supplier No. BS12911RCY3
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Phospho-c-Fos (Ser374) Polyclonal antibody specifically detects Phospho-c-Fos (Ser374) in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)

Specifications

Antigen Phospho-c-Fos (Ser374)
Applications Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Cyanine3
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene FOS
Gene Accession No. P01100, P01101, P12841
Gene Alias activator protein 1; AP-1; cellular oncogene c-fos; cellular oncogene fos; cFos; C-FOS; c-fos oncogene; c-fos protooncogene protein; D12Rfj1; FBJ murine osteosarcoma viral (v-fos) oncogene homolog (oncogene FOS); FBJ murine osteosarcoma viral oncogene homolog; FBJ murine osteosarcoma viral oncogene homolog B; FBJ osteosarcoma oncogene; FBJ osteosarcoma oncogene B; FBJ osteosarcoma viral oncogene isoform deltaFosb-2; FOS; Fos proto-oncogene, AP-1 trancription factor subunit; Fos proto-oncogene, AP-1 transcription factor subunit; Fosb; FosB proto-oncogene, AP-1 transcription factor subunit; fra-2; G0/G1 switch regulatory protein 7; G0S7; I79_009428; p55; protein fosB; proto-oncogene cFOS; proto-oncogene c-Fos; proto-oncogene protein c-fos; v-fos FBJ murine osteosarcoma viral oncogene homolog
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human c-Fos around the phosphorylation site of Ser374
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14281, 2353, 314322
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.