Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-c-Fos (Ser374) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564675
Description
Phospho-c-Fos (Ser374) Polyclonal Antibody detects endogenous levels of c-Fos only when phosphorylated at Ser374.
FBJ murine osteosarcoma viral oncogene homolog is commonly known as Cellular protooncogene Fos (c-Fos). It belongs to the fos transcription factor family and is encoded by the C-FOS gene located on chromosome 14. c-Fos heterodimerizes with c-Jun to form the Activator Protein -1 (AP-1) transcription factor which regulates transcription of several genes that play an important role in cellular signal transduction, cell proliferation and differentiation. c-Fos has been shown to be phosphorylated by various stimuli including growth factors and insulin at atleast seven different sites. The best known upstream kinase for phosphorylation at Threonine 232 is ERK MAPK which regulates localization of c-Fos to the nucleus and is thus important for c-Fos induced transcriptional activity.
Specifications
Phospho-c-Fos (Ser374) | |
Polyclonal | |
Unconjugated | |
FOS | |
activator protein 1; AP-1; cellular oncogene c-fos; cellular oncogene fos; cFos; C-FOS; c-fos oncogene; c-fos protooncogene protein; D12Rfj1; FBJ murine osteosarcoma viral (v-fos) oncogene homolog (oncogene FOS); FBJ murine osteosarcoma viral oncogene homolog; FBJ murine osteosarcoma viral oncogene homolog B; FBJ osteosarcoma oncogene; FBJ osteosarcoma oncogene B; FBJ osteosarcoma viral oncogene isoform deltaFosb-2; FOS; Fos proto-oncogene, AP-1 trancription factor subunit; Fos proto-oncogene, AP-1 transcription factor subunit; Fosb; FosB proto-oncogene, AP-1 transcription factor subunit; fra-2; G0/G1 switch regulatory protein 7; G0S7; I79_009428; p55; protein fosB; proto-oncogene cFOS; proto-oncogene c-Fos; proto-oncogene protein c-fos; v-fos FBJ murine osteosarcoma viral oncogene homolog | |
Rabbit | |
Affinity Chromatography | |
RUO | |
14281, 2353, 314322 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P01100, P01101, P12841 | |
FOS | |
A synthesized peptide derived from human Fos (Phospho-Ser374). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction