Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-c-Jun (Thr93) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104750
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104750 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104750 Supplier Invitrogen™ Supplier No. PA5104750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of c-Jun only when phosphorylated at Threonine 93.

c-Jun is a transcription factor that recognizes and binds to the enhancer heptamer motif 5'-TGA[CG]TCA-3'. It promotes activity of NR5A1 when phosphorylated by HIPK3 leading to increased steroidogenic gene expression upon cAMP signaling pathway stimulation. Involved in activated KRAS-mediated transcriptional activation of USP28 in colorectal cancer (CRC) cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-c-Jun (Thr93)
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene JUN
Gene Accession No. P05412, P05627, P17325
Gene Alias Activator protein 1; AH119; AP1; AP-1; Avian sarcoma virus 17 (v-jun) oncogene homolog; cjun; c-Jun; c-jun transcription factor; enhancer-binding protein AP1; I79_009953; immediate early; JUN; Jun A; Jun activation domain binding protein; Jun oncogene; jun proto-oncogene; Jun proto-oncogene, AP-1 transcription factor subunit; Junc; JUND; p39; proto-oncogene c-Jun; Rjg-9; transcription factor; transcription factor AP-1; V-jun avian sarcoma virus 17 oncogene homolog; v-jun sarcoma virus 17 oncogene homolog
Gene Symbols JUN
Host Species Rabbit
Immunogen A synthesized peptide derived from human JUN(Accession P05412), corresponding to amino acid residues around phosphorylated Thr93.
Purification Method Sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16476, 24516, 3725
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.