Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospho-c-Myc (Ser347) Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABC146125UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC146125UL 25 μL
ABC1461100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABC146125UL Supplier MilliporeSigma Supplier No. ABC146125UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

phospho-c-Myc (Ser347) Polyclonal antibody specifically detects phospho-c-Myc (Ser347) in Human samples. It is validated for Peptide Inhibition Assay, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-c-Myc (Ser347)
Applications Inhibition Assays, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 10 amino acids surrounding phosphoserine 347 from the C-terminal half of human c-Myc.
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for 1 year at +2°C to +8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.