Learn More
phospho-c-Myc (Ser348) Rabbit anti-Human, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | phospho-c-Myc (Ser348) |
| Applications | Functional Assay, Inhibition Assays, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated linear peptide corresponding to 10 amino acids from surrounding phosphoserine 348 from the C-terminal half of human c-Myc. |
| Purification Method | Affinity Purified |
| Quantity | 25 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.