Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospho-c-Myc (Ser81) Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABC147125UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC147125UG 25 μg
ABC1471100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABC147125UG Supplier MilliporeSigma Supplier No. ABC147125UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

phospho-c-Myc (Ser81) Polyclonal antibody specifically detects phospho-c-Myc (Ser81) in Human samples. It is validated for Affects Function, Peptide Inhibition Assay, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-c-Myc (Ser81)
Applications Functional Assay, Inhibition Assays, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 10 amino acids surrounding phosphoserine 81 from human c-Myc.
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for 1 year at +2°C to +8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.