Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-c-Raf (Ser338) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985353
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5353 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5353 Supplier Bioss Supplier No. BS3377R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Raf-1 (c-Raf) is a MAP kinase kinase kinase (MAP3K) which functions downstream of the Ras family of membrane associated GTPases to which it binds directly. Once activated Raf-1 can phosphorylate to activate the dual specificity protein kinases MEK1 and MEK2 which in turn phosphorylate to activate the serine/threonine specific protein kinases ERK1 and ERK2. Activated ERKs are pleiotropic effectors of cell physiology and play an important role in the control of gene expression involved in the cell division cycle, apoptosis, cell differentiation and cell migration. Mutations in this gene are associated with Noonan syndrome 5 and LEOPARD syndrome 2.

Specifications

Antigen Phospho-c-Raf (Ser338)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Raf1
Gene Accession No. P04049
Gene Alias 6430402F14Rik; AA990557; BB129353; CMD1NN; Craf; c-Raf; C-Raf proto-oncogene, serine/threonine kinase; Craf1; Craf1 transforming; D830050J10Rik; EC 2.7.11.1; kinase Raf1; murine leukemia viral (v-raf-1) oncogene homolog 1 (3611-MSV); murine sarcoma 3611 oncogene 1; NS5; Oncogene RAF1; OTTHUMP00000207813; OTTHUMP00000209389; Proto-oncogene c-RAF; RAF; raf proto-oncogene serine/threonine protein kinase; RAF proto-oncogene serine/threonine-protein kinase; RAF1; Raf-1; Raf-1 proto-oncogene, serine/threonine kinase; sarcoma 3611 oncogene; v-Raf; v-raf-1 leukemia viral oncogene 1; vraf1 murine le; v-raf-1 murine leukemia viral oncogene homolog 1; v-raf-1 murine leukemia viral oncogene-like protein 1; v-raf-leukemia viral oncogene 1
Gene Symbols Raf1
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human c-Raf around the phosphorylation site of Ser338.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5894
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.