Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-C2orf33 (Ser172, Ser146) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104614
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104614 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104614 Supplier Invitrogen™ Supplier No. PA5104614
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of MFF only when phosphorylated at Ser172/146.

Plays a role in mitochondrial and peroxisomal fission. Promotes the recruitment and association of the fission mediator dynamin-related protein 1 (DNM1L) to the mitochondrial surface. May be involved in regulation of synaptic vesicle membrane dynamics by recruitment of DNM1L to clathrin-containing vesicles. [Uniprot].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-C2orf33 (Ser172, Ser146)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MFF
Gene Accession No. Q9GZY8
Gene Alias 5230400G24Rik; AD030; AD033; AI314724; C2H2orf33; C2orf33; GL004; hypothetical protein LOC393508; hypothetical protein LOC506291; MFF; mffa; Mitochondrial fission factor; mitochondrial fission factor a; mitochondrial fission factor homolog B; RGD1310230; zgc:66022
Gene Symbols MFF
Host Species Rabbit
Immunogen A synthesized peptide derived from human MFF(Accession Q9GZY8), corresponding to amino acid residues around phosphorylated Ser172.
Purification Method Sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56947
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.