Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-CaMK2 (Thr287) Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS1647R

Description
The Ca2+/calmodulin-dependent protein kinases (CaM kinases) comprise a structurally related subfamily of serine/threonine kinases which include CaMKI, CaMKII and CaMKIV. CaMKII is an ubiquitously expressed serine/threonine protein kinase that is activated by Ca2+/calmodulin (CaM) binding and autophosphorylation and has been implicated in regulation of the cell cycle and transcription. It is involved in dendritic spine and synapse formation, neuronal plasticity and regulation of sarcoplasmic reticulum Ca2+ transport in skeletal muscle. In neurons, plays an essential structural role in the reorganization of the actin cytoskeleton during plasticity by binding and bundling actin filaments in a kinase-independent manner.Specifications
Phospho-CaMK2 (Thr287) | |
Polyclonal | |
Unconjugated | |
CAMK2A | |
alpha CaM kinase II; alpha-CaMKII; Ca2+/calmodulin-dependent protein kinase II; Ca2+/calmodulin-dependent protein kinase II alpha; Calci; calcium/calmodulin dependent protein kinase II alpha; calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha; calcium/calmodulin-dependent protein kinase II alpha; calcium/calmodulin-dependent protein kinase II alpha subunit; calcium/calmodulin-dependent protein kinase II alpha-B subunit; calcium/calmodulin-dependent protein kinase type II alpha chain; Calcium/calmodulin-dependent protein kinase type II subunit alpha; CaM kinase II alpha subunit; CaM kinase II subunit alpha; CaMK II; CAMK2; Camk2a; CAMKA; CaMKII; CaMK-II alpha subunit; caMK-II subunit alpha; CaMKIINalpha; caM-kinase II alpha chain; EC 2.7.11.17; KIAA0968; mKIAA0968; OTTHUMP00000165787; OTTHUMP00000165788; Phospho-CaMKI; PK2CDD; PKCCD; R74975 | |
Rabbit | |
Protein A | |
RUO | |
815 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9UQM7 | |
CAMK2A | |
KLH conjugated synthetic phosphopeptide derived from mouse CaMK2 alpha around the phosphorylation site of Thr287. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction