Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-CaMKI (Thr177) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105644
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105644 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105644 Supplier Invitrogen™ Supplier No. PA5105644
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of CaMK1 alpha only when phosphorylated at Thr177.

CAMK1 (CaMK1) is a serine/threonine kinase that is a member of the calcium/calmodulin-dependent protein kinase family. It is involved in cell differentiation, nervous system development, and signal transduction. CAMK1 is expressed in many tissues and is a component of a calmodulin-dependent protein kinase cascade. Calcium/calmodulin directly activates calcium/calmodulin-dependent protein kinase I by binding to the enzyme and indirectly promotes the phosphorylation and synergistic activation of the enzyme by calcium/calmodulin-dependent protein kinase I kinase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-CaMKI (Thr177)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CAMK1
Gene Accession No. Q14012, Q63450, Q91YS8
Gene Alias AI505105; calcium/calmodulin dependent protein kinase I; calcium/calmodulin-dependent protein kinase I; calcium/calmodulin-dependent protein kinase type 1; CaM kinase I; CaM kinase I alpha; Camk1; CAMK1-PEN; Camki; caM-KI; CaMKIalpha; CaMKI-alpha; D6Ertd263e; G alpha interacting protein; Gaip; MGC120317; MGC120318; regulator of G-protein signalling 19
Gene Symbols CAMK1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CAMK1(Accession Q14012), corresponding to amino acid residues around phosphorylated Thr177.
Purification Method sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171503, 52163, 8536
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.