Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-CaMKII alpha/beta/delta (Tyr231) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA517108
Description
It is not recommended to aliquot this antibody.
The Ca2+/calmodulin-dependent protein kinases (CaM kinases) comprise a structurally related subfamily of serine/threonine kinases which include CaMKI, CaMKII and CaMKIV. CaMKII is an ubiquitously expressed serine/threonine protein kinase that is activated by Ca2+ and calmodulin (CaM) and has been implicated in regulation of the cell cycle and transcription. There are four CaMKII isozymes, designated alpha, beta, gamma and delta, which may or may not be co-expressed in the same tissue type. CaMKII alpha is autophosphorylated on Thr 286 upon the binding of the Ca2+/CaM complex to the autoinhibitory domain of CaMKII. This process is called Ca2+/CaM trapping, which is thought to be involved in the synaptic encoding of information.
Specifications
| Phospho-CaMKII alpha/beta/delta (Tyr231) | |
| Polyclonal | |
| 0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
| P08413, P11275, P15791, Q13554, Q13557, Q9UQM7 | |
| CAMK2A, CAMK2B, CAMK2D | |
| Synthetic phosphopeptide corresponding to residues surrounding pTyr231 of human CaMKII | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| CAMK2B | |
| [d]-CaMKII; 2810011D23Rik; 8030469K03Rik; alpha CaM kinase II; alpha-CaMKII; Ca++/calmodulin-dependent protein kinase 2 delta subunit; Ca++/calmodulin-dependent protein kinase II beta subunit; Ca++/calmodulin-dependent protein kinase II delta subunit; Ca++/calmodulin-dependent protein kinase II, delta subunit; Ca2+/calmodulin-dependent protein kinase II alpha; Calci; calcium/calmodulin dependent protein kinase II alpha; calcium/calmodulin dependent protein kinase II beta; calcium/calmodulin dependent protein kinase II delta; calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha; calcium/calmodulin-dependent protein kinase (CaM kinase) II beta; calcium/calmodulin-dependent protein kinase (CaM kinase) II delta; calcium/calmodulin-dependent protein kinase II alpha; calcium/calmodulin-dependent protein kinase II alpha subunit; calcium/calmodulin-dependent protein kinase II alpha-B subunit; calcium/calmodulin-dependent protein kinase II beta; calcium/calmodulin-dependent protein kinase II delta; calcium/calmodulin-dependent protein kinase II, beta; calcium/calmodulin-dependent protein kinase II, delta; calcium/calmodulin-dependent protein kinase type II alpha chain; calcium/calmodulin-dependent protein kinase type II beta chain; calcium/calmodulin-dependent protein kinase type II delta chain; calcium/calmodulin-dependent protein kinase type II subunit alpha; Calcium/calmodulin-dependent protein kinase type II subunit beta; calcium/calmodulin-dependent protein kinase type II subunit delta; CaM kinase II alpha subunit; CaM kinase II beta subunit; CaM kinase II delta subunit; caM kinase II subunit alpha; CaM kinase II subunit beta; caM kinase II subunit delta; CAM2; CAMK; CaMK II; CAMK1; Camk2; CaMK2 delta; CaMK-2 delta; CaMK2- delta; Camk2a; CaMK-2a; CaMK2-a; Camk2b; Camk2d; CAMK2G; CAMKA; CAMKB; CAMKD; CAMKG; Camki; CaMKII; CAMK-II; CaMK-II alpha subunit; CaMK-II delta subunit; caMK-II subunit alpha; caMK-II subunit beta; caMK-II subunit delta; CaMKIINalpha; caM-kinase II alpha chain; CaM-kinase II beta chain; CaM-kinase II delta chain; Ck2b; KIAA0968; Kiaa4163; mKIAA0968; PK2CDD; PKCCD; proline rich calmodulin-dependent protein kinase; R74975; RATCAMKI | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 24245, 24246, 25400, 815, 816, 817 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction