Learn More
Phospho-CamKII (Thr286) Polyclonal Antibody, Abbomax
Rabbit Polyclonal Antibody
Supplier: Abbomax Inc 600270

Description
Positive control: Human thyroid carcinoma, cardiac and skeletal muscle tissues Cellular location: Cytoplasm.
Phospho-CamKII (Thr286) Polyclonal antibody specifically detects Phospho-CamKII (Thr286) in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
Phospho-CamKII (Thr286) | |
Polyclonal | |
Unconjugated | |
CAMK2B | |
[d]-CaMKII; 2810011D23Rik; 8030469K03Rik; alpha CaM kinase II; alpha-CaMKII; Ca++/calmodulin-dependent protein kinase 2 delta subunit; Ca++/calmodulin-dependent protein kinase II beta subunit; Ca++/calmodulin-dependent protein kinase II delta subunit; Ca++/calmodulin-dependent protein kinase II, delta subunit; Ca2+/calmodulin-dependent protein kinase II alpha; Calci; calcium/calmodulin dependent protein kinase II alpha; calcium/calmodulin dependent protein kinase II beta; calcium/calmodulin dependent protein kinase II delta; calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha; calcium/calmodulin-dependent protein kinase (CaM kinase) II beta; calcium/calmodulin-dependent protein kinase (CaM kinase) II delta; calcium/calmodulin-dependent protein kinase II alpha; calcium/calmodulin-dependent protein kinase II alpha subunit; calcium/calmodulin-dependent protein kinase II alpha-B subunit; calcium/calmodulin-dependent protein kinase II beta; calcium/calmodulin-dependent protein kinase II delta; calcium/calmodulin-dependent protein kinase II, beta; calcium/calmodulin-dependent protein kinase II, delta; calcium/calmodulin-dependent protein kinase type II alpha chain; calcium/calmodulin-dependent protein kinase type II beta chain; calcium/calmodulin-dependent protein kinase type II delta chain; calcium/calmodulin-dependent protein kinase type II subunit alpha; Calcium/calmodulin-dependent protein kinase type II subunit beta; calcium/calmodulin-dependent protein kinase type II subunit delta; CaM kinase II alpha subunit; CaM kinase II beta subunit; CaM kinase II delta subunit; caM kinase II subunit alpha; CaM kinase II subunit beta; caM kinase II subunit delta; CAM2; CAMK; CaMK II; CAMK1; Camk2; CaMK2 delta; CaMK-2 delta; CaMK2- delta; Camk2a; CaMK-2a; CaMK2-a; Camk2b; Camk2d; CAMK2G; CAMKA; CAMKB; CAMKD; CAMKG; Camki; CaMKII; CAMK-II; CaMK-II alpha subunit; CaMK-II delta subunit; caMK-II subunit alpha; caMK-II subunit beta; caMK-II subunit delta; CaMKIINalpha; caM-kinase II alpha chain; CaM-kinase II beta chain; CaM-kinase II delta chain; Ck2b; KIAA0968; Kiaa4163; mKIAA0968; PK2CDD; PKCCD; proline rich calmodulin-dependent protein kinase; R74975; RATCAMKI | |
A synthetic peptide corresponding to the epitope with a single phosphorylation site Thr 286 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
25mM tris with proprietary stabilizer and 0.01% sodium azide | |
Q13554, Q13557, Q9UQM7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
815, 816, 817 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.