Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-CaMKIV (Thr196, Thr200) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538435
Description
Ca2+/calmodulin dependent protein kinase type IV (CAMKIV) belongs to the serine/threonine protein kinase family, and to the Ca(2+)/calmodulin-dependent protein kinase subfamily. This enzyme is a multifunctional serine/threonine protein kinase with limited tissue distribution, that has been implicated in transcriptional regulation in lymphocytes, neurons and male germ cells.
Specifications
Phospho-CaMKIV (Thr196, Thr200) | |
Polyclonal | |
Unconjugated | |
CAMK4 | |
Calcium/calmodulin-dependent protein kinase type IV, CaMK IV, CaM kinase-GR, CAMK4, CAMK, CAMK-GR, CAMKIV | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12326, 25050, 814 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q16566, P08414, P13234 | |
CAMK4 | |
A synthetic phosphopeptide derived from human CaMK4 around the phosphorylation site of Thr196/200 (M-K-TP-V-C) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction