Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Casein kinase 1 isoforms gamma 1/2/3 (Tyr263) Rabbit anti-Human∣Mouse∣Rat, Polyclonal, Bioss
SDP

Catalog No. 501982233
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2233 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2233 Supplier Bioss Supplier No. BS12426R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Casein kinases are operationally defined by their preferential utilization of acidic proteins such as caseins as substrates. They can phosphorylate a large number of proteins. Participates in Wnt signaling.

Specifications

Antigen Phospho-Casein kinase 1 isoforms gamma 1/2/3 (Tyr263)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CSNK1G1
Gene Accession No. P78368, Q62761, Q62762, Q62763, Q8BTH8, Q8BVP5, Q8C4X2, Q9HCP0, Q9Y6M4
Gene Symbols CSNK1G1, CSNK1G2, CSNK1G3
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human Casein kinase I isoforms gamma 1+2+3 around the phosphorylation site of Tyr263.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103236, 1455, 1456, 214897, 53944, 64086, 64823, 65278, 70425
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.