Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Caspase 2 (Ser157) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538566
Description
This gene encodes a protein which is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. The proteolytic cleavage of this protein is induced by a variety of apoptotic stimuli. Alternative splicing of this gene results in multiple transcript variants that encode different isoforms.
Specifications
Phospho-Caspase 2 (Ser157) | |
Polyclonal | |
Unconjugated | |
CASP2 | |
CASP-2, ICH1, NEDD-2, NEDD2, PPP1R57, caspase-2, neural precursor cell expressed developmentally down-regulated protein 2, protease ICH-1, protein phosphatase 1, regulatory subunit 57 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
835 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P42575 | |
CASP2 | |
A synthetic phosphopeptide derived from human Caspase 2 around the phosphorylation site of Ser157 (R-L-SP-T-D) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction