Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-CCR5 (Ser349) Chimeric Recombinant Rabbit Monoclonal Antibody (E11/19)
GREENER_CHOICE

Catalog No. PIMA548293
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA548293 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA548293 Supplier Invitrogen™ Supplier No. MA548293
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Specificity: The E11/19 monoclonal antibody binds to phosphoserine 349 corresponding to 22 c-terminal amino acid residue of human CCR5. CCR5 is a receptor for a number of inflammatory CC-chemokines including CCL3/MIP-1-alpha, CCL4/MIP-1-beta and RANTES.

Human immunodeficiency virus (HIV) and related virus require coreceptors, in addition to CD4, to infect target cells. Some G protein-coupled receptors including CCR5, CXCR4, CCR3, CCR2b and CCR8 in the chemokine receptor family, and four new human molecules GPR15, STRL33, GPR1 and V28 were recently identified as HIV coreceptors. Among them, CCR5 (CC-CKR-5) is a principal coreceptor for macrophage- and dual-tropic HIV-1 strains fusion and entry of human white blood cells. CCR5 is required for the infection by HIV-1, HIV-2, and SIV. The beta-chemokines RANTES, MIP-alpha and MIP-beta are the ligands for CCR5 and prevent infection by M-tropic HIV-1. CCR5 associates with the surface CD4-gp120 of HIV complex and leads to membrane fusion and virus entry of target cells. The amino-terminal domain and the extracellular loops of CCR5 serve as HIV biding sites. CCR5 messenger RNA is expressed in lymphoid organs and monocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-CCR5 (Ser349) Chimeric
Applications ELISA, Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone E11/19
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene CCR5
Gene Accession No. P51681
Gene Alias AM4-7; C-C chemokine receptor type 5; C-C CKR-5; C-C motif chemokine receptor 5 (gene/pseudogene); C-C motif chemokine receptor 5 A159A; CCCKR5; CC-CKR-5; Ccr5; CCR-5; CD195; CD195 antigen; chemokine (C-C motif) receptor 5; chemokine (C-C) receptor 5; chemokine receptor CCR5; chemr13; CKR5; CKR-5; Cmkbr5; HIV-1 fusion coreceptor; IDDM22; MIP-1 alpha receptor
Gene Symbols CCR5
Host Species Rabbit
Immunogen This antibody was raised by immunizing mice with a synthetic phosphopeptide (CEAPERA(pS)(pS)VYTR(pS)TGEQI(pS)VGL) corresponding to the C-terminal region of human CCR5.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1234
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.