Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-CD104 Integrin beta 4 (Tyr1526) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143712
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143712 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143712 Supplier Invitrogen™ Supplier No. PA5143712
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody was cross-adsorbed to phospho-tyrosine coupled to agarose and to unphosphorylated Integrin beta4 (Tyr-1526) peptide before affinity purification using phospho-Integrin beta4 (Tyr-1526) peptide (without carrier). The purified antibody detects a 200kDa* protein corresponding to the molecular mass of Integrin beta4 on SDS-PAGE immunoblots of A431 and src-transformed Hct116 cells treated with pervanadate, but not in control cells.

Integrins are heterodimers comprised of alpha and beta subunits, that are noncovalently associated transmembrane glycoprotein receptors. Different combinations of alpha and beta polypeptides form complexes that vary in their ligand-binding specificities. Integrins mediate cell-matrix or cell-cell adhesion, and transduced signals that regulate gene expression and cell growth. This gene encodes the integrin beta 4 subunit, a receptor for the laminins. This subunit tends to associate with alpha 6 subunit and is likely to play a pivotal role in the biology of invasive carcinoma. Mutations in this gene are associated with epidermolysis bullosa with pyloric atresia. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-CD104 Integrin beta 4 (Tyr1526)
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene ITGB4
Gene Accession No. A2A863, P16144, Q64632
Gene Alias AA407042; C230078O20; CD antigen CD104; CD104; CD104 antigen; GP150; integrin beta 4; integrin beta-4; integrin subunit beta 4; integrin, beta 4; Itgb4
Gene Symbols ITGB4
Host Species Rabbit
Immunogen Phospho-Integrin beta4 (Tyr-1526) synthetic peptide (coupled to KLH) corresponding to amino acid residues around tyrosine 1526 of human Integrin beta4. This peptide sequence is found in all three Integrin beta4 isoforms and is highly conserved in rat and mouse Integrin beta4.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192897, 25724, 3691
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.