Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-CD136 (RON) (Tyr1238) Polyclonal Antibody, Abbomax

Catalog No. 502204021
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4021 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4021 Supplier Abbomax Inc Supplier No. 601910
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Human stomach tissue Cellular location: Membrane.

Phospho-CD136 (RON) (Tyr1238) Polyclonal antibody specifically detects Phospho-CD136 (RON) (Tyr1238) in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Phospho-CD136 (RON) (Tyr1238)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation 25mM tris with proprietary stabilizer and 0.01% sodium azide
Gene MST1R
Gene Accession No. Q04912
Gene Alias CD136; Cdw136; c-met-related tyrosine kinase; friend virus susceptibility 2; Fv2; Fv-2; macrophage stimulating 1 receptor; macrophage stimulating 1 receptor (c-met-related tyrosine kinase); macrophage-stimulating protein receptor; Macrophage-stimulating protein receptor alpha chain; Macrophage-stimulating protein receptor beta chain; MSP receptor; Mst1r; MST1R variant RON30; MST1R variant RON62; p185-Ron; protein tyrosine kinase 8; Protein-tyrosine kinase 8; PTK8; PTK8 protein tyrosine kinase 8; receptor protein tyrosine kinase, c-met-related; RON; RON variant E2E3; Stem cell-derived tyrosine kinase; STK
Host Species Rabbit
Immunogen A synthetic peptide containing epitope -EPYYS- with a single phosphorylation sites Y1238 of the human CD136
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4486
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.