Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-CD247 (CD3 zeta) (Lys142) Monoclonal Antibody (EM-54), Alexa Fluor™ 647
GREENER_CHOICE

Catalog No. PIMA538687
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538687 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA538687 Supplier Invitrogen™ Supplier No. MA538687
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

This antibody recognizes phosphorylated intracellular tyrosine 142 of CD3 zeta chain (CD247), which is a component of TCR/CD3 complex expressed on T cells.

CD3z complex is crucial in transducing antigen-recognition signals into the cytoplasm of T cells and in regulating the cell surface expression of the TCR complex. T cell activation through the antigen receptor (TCR) involves the cytoplasmic tails of the CD3 subunits CD3 gamma, CD3 delta, CD3 epsilon and CD3 zeta. These CD3 subunits are structurally related members of the immunoglobulins super family encoded by closely linked genes on human chromosome 11. The CD3 components have long cytoplasmic tails that associate with cytoplasmic signal transduction molecules, and this association is mediated at least in part by a double tyrosine-based motif present in a single copy in the CD3 subunits. CD3 may play a role in TCR-induced growth arrest, cell survival and proliferation. The CD3 antigen is present on 68-82% of normal peripheral blood lymphocytes, 65-85% of thymocytes and Purkinje cells in the cerebellum. It is never expressed on B or NK cells. Decreased percentages of T lymphocytes may be observed in some autoimmune diseases. Diseases associated with CD3z include Immunodeficiency 25 and T-B+ Severe Combined Immunodeficiency Due To Cd3delta/Cd3epsilon/Cd3zeta.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-CD247 (CD3 zeta) (Lys142)
Applications Flow Cytometry
Classification Monoclonal
Clone EM-54
Concentration 100 μg/mL
Conjugate Alexa Fluor 647
Formulation PBS with 0.2% BSA and 15mM sodium azide
Gene CD247
Gene Accession No. P20963, P24161
Gene Alias 4930549J05Rik; A430104F18Rik; antigen CD3Z, zeta polypeptide; AW552088; Cd247; CD247 antigen; CD247 antigen, zeta subunit; Cd247 molecule; Cd3; CD3 antigen, zeta polypeptide; CD3 zeta; CD3 zeta chain; CD3e antigen; CD3-epsilon; Cd3-eta; CD3H; CD3Q; Cd3z; CD3Z antigen, zeta polypeptide (TiT3 complex); Cd3zeta; Cd3-zeta; CD3zeta chain; CD3-zeta/eta; FLJ18683; IMD25; T3E; T3z; T-cell antigen receptor complex, zeta subunit of CD3; T-cell receptor CD3 subunit zeta; T-cell receptor CD3, subunit zeta; T-cell receptor T3 eta chain; T-cell receptor T3 zeta chain; T-cell receptor zeta chain; T-cell surface glycoprotein CD3 zeta chain; TCR zeta chain; TCRE; Tcrk; TCRZ; TCRzeta
Gene Symbols CD247
Host Species Mouse
Immunogen CP1 (phosphorylated Tyr142) - KLH.
Purification Method Size-exclusion chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12503, 919
Target Species Human, Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.