Learn More
Invitrogen™ Phospho-Cdc6 (Ser54) Recombinant Rabbit Monoclonal Antibody (0K9Y2)

Description
Positive Samples: NIH/3T3 Immunogen sequence: PLSPR.
Specifications
Specifications
| Antigen | Phospho-Cdc6 (Ser54) |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 0K9Y2 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CDC6 |
| Gene Accession No. | O89033 |
| Gene Alias | CDC18(S.pombe); CDC18L; cdc18-related protein; CDC6; CDC6 cell division cycle 6 homolog; CDC6-related protein; Cell division control protein 6 homolog; cell division cycle 6; cell division cycle 6 homolog; HsCDC18; HsCDC6; p62(cdc6) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.