Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-CDCP1 (Tyr707) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5105230
Description
Antibody detects endogenous levels of CDCP1 only when phosphorylated at Tyr707.
CDCP1 is a transmembrane protein containing three extracellular CUB domains. It is found to be overexpressed in colon and lung cancers. Its expression level is correlated with the metastatic ability of carcinoma cells. CDCP1 is located on the cell surface. It has been shown to be tyrosine phosphorylated in a cancer cell line. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications
Phospho-CDCP1 (Tyr707) | |
Polyclonal | |
Unconjugated | |
Cdcp1 | |
9030022E12Rik; AA409659; CD318; CDCP1; CUB domain containing protein 1; CUB domain-containing protein 1; E030027H19Rik; gp140; Membrane glycoprotein gp140; RGD1305578; SIMA135; Subtractive immunization M plus HEp3-associated 135 kDa protein; transmembrane and associated with src kinases; TRASK; UNQ2486/PRO5773 | |
Rabbit | |
Sequential Chromatography | |
RUO | |
109332, 301082, 64866 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q5U462, Q9H5V8 | |
Cdcp1 | |
A synthesized peptide derived from human CDCP1(Accession Q9H5V8), corresponding to amino acid residues around phosphorylated Tyr707. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction