Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-CDK1/CDK2 (Thr14) Recombinant Rabbit Monoclonal Antibody (6R1O7)
GREENER_CHOICE

Catalog No. PIMA535854
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535854 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535854 Supplier Invitrogen™ Supplier No. MA535854
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: EGTYG.

Voltage dependent anion channel required for acidification and functions of the Golgi apparatus that may function in counter-ion conductance.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-CDK1/CDK2 (Thr14)
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 6R1O7
Concentration 1.04 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CDK2
Gene Accession No. P06493, P11440, P24941, P39951, P97377, Q63699
Gene Alias A630093N05Rik; Cdc2; CDC28A; Cdc2a; cdc2-related protein kinase; Cdk1; CDK2; CDKN1; CDKN2; cell cycle controller CDC2; cell cycle p34 CDC2 kinase protein; cell division control protein 2 homolog; cell division cycle 2 homolog A; cell division cycle 2, G1 to S and G2 to M; Cell division cycle control protein 2; cell division cycle control protein 2a; cell division protein kinase 1; Cell division protein kinase 2; cyclin dependent kinase 1; cyclin dependent kinase 2; cyclin dependent kinase 2-alpha; cyclin-dependent kinase 1; Cyclin-dependent kinase 2; p33 protein kinase; p33(CDK2); p34 protein kinase; p34>CDC2>; P34CDC2
Gene Symbols CDK1, CDK2
Host Species Rabbit
Immunogen A synthetic phosphorylated peptide around T14 of human Cdk1/2CDK1 (P06493).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1017, 12534, 12566, 362817, 54237, 983
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.