Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-CDK1 (Tyr15) Recombinant Rabbit Monoclonal Antibody (BLR101H)

Rabbit Recombinant Monoclonal Antibody
$173.10 - $436.00
Specifications
| Antigen | Phospho-CDK1 (Tyr15) |
|---|---|
| Clone | BLR101H |
| Concentration | 40 μg/mL |
| Content And Storage | 4°C |
| Applications | Immunoprecipitation, Western Blot |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA544393
![]() |
Invitrogen™
MA544393 |
100 μL |
Each for $436.00
|
|
|||||
|
PIMA544394
|
Invitrogen™
MA544394 |
20 μL |
Each for $173.10
|
|
|||||
Description
Phospho-CDK1 (Tyr15) Recombinant Monoclonal Antibody for Western Blot, IP
CDK1 or Cell Division Control protein 1 is essential for the completion of START, the controlling event in the cell cycle that is required to initiate mitosis. CDK1 is a catalytic subunit of a protein kinase complex, called the M-Phase Promoting Factor that induces entry into mitosis and is universal among eukaryotes. Phosphorylation of Bcl-2 in G2/M phase-arrested cells following photodynamic therapy with hypericin involves a CDK1-mediated signal and delays the onset of apoptosis. Therapeutic potential of CDK inhibitor NU2058 in androgen-independent prostate cancer has also been demonstrated.Specifications
| Phospho-CDK1 (Tyr15) | |
| 40 μg/mL | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| BBS with 0.1% BSA and 0.09% sodium azide; pH 7.8-8.6 | |
| P06493 | |
| 983 | |
| Peptide surrounding phosphorylated tyrosine 15 of human cyclin dependent kinase 1. | |
| Antibody |
| BLR101H | |
| 4°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| CDK1 | |
| 5363; ABL1; cdc; CDC2; cdc-2; cdc2 kinase; CDC28A; Cdc2a; cdc2-a; cdc2Dm; cdc2x1.1; CDCDm; CDK1; Cdk-1; cdk1.S; CDK1/CDC2; CDK1-A; cdk1-b; Cdk1-PA; CDKN1; cell cycle controller CDC2; cell cycle p34 CDC2 kinase protein; cell division control; cell division control protein 2 homolog; Cell division control protein 2 homolog 1; cell division control protein 2-A; cell division cycle 2 homolog A; cell division cycle 2 like; cell division cycle 2, G1 to S and G2 to M; Cell division cycle control protein 2; cell division cycle control protein 2a; cell division protein kinase 1; Cell division protein kinase 1-A; CG5363; CG5363-PA; cyclin dependent kinase; cyclin dependent kinase 1; cyclin-dependent kinase; Cyclin-dependent kinase 1; cyclin-dependent kinase 1 S homeolog; cyclin-dependent kinase 1-A; D2366; Dcdc2; dCdk1; DKFZp686L20222; Dm cdc2; Dmcdc2; DmCdk1; Dmel\CG5363; Dmel_CG5363; DNA polymerase delta catalytic subunit; DNA polymerase III; DNA-directed DNA polymerase delta POL3; group 4; HPR6; KCIP-1; l(2)31Eh; LD38718p; maturation-promoting factor; MGC111195; MGC53829; MPF; OTTHUMP00000019660; P34 CDK1; P34 MGC111195; p34 protein kinase; p34 protein kinase 1; p34>CDC2>; p34cdc2; p34-cdc2 homologue; POL3; Protein 1054; protein 2; PSTAIR; TEX1; unnamed protein product; xcdc2; XELAEV_18036802mg; YDL102W | |
| CDK1 | |
| Primary | |
| Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title