Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-CDK20 (Thr161) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105308
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105308 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105308 Supplier Invitrogen™ Supplier No. PA5105308
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of CDK20 only when phosphorylated at Thr161.

The protein encoded by this gene contains a kinase domain most closely related to the cyclin-dependent protein kinases. The encoded kinase may activate cyclin-dependent kinase 2 and is involved in cell growth. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-CDK20 (Thr161)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CDK20
Gene Accession No. Q4KM34, Q8IZL9, Q9JHU3
Gene Alias 4932702G04Rik; AU019450; AW558027; CAK-kinase p42; Ccrk; CDCH; CDK20; CDK-activating kinase p42; CDK-related protein kinase PNQLARE; cell cycle related kinase; cell cycle-related kinase; Cell division protein kinase 20; cyclin dependent kinase 20; cyclin-dependent kinase 20; Cyclin-dependent protein kinase H; cyclin-kinase-activating kinase p42; p42; PNQALRE; PNQLARE
Gene Symbols CDK20
Host Species Rabbit
Immunogen A synthesized peptide derived from human CDK20(Accession Q8IZL9), corresponding to amino acid residues around phosphorylated Thr161.
Purification Method Sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 105278, 23552, 364666
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.