Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-CDK6 (Tyr24) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986464
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6464 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6464 Supplier Bioss Supplier No. BS5287R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cyclin-dependent kinase 6 (p40cdk6 also known as PLSTIRE) belongs to a family of proteins designated as cdks. They are critical regulators of cell cycle progression. The prototype member of this family, p34cdc2, and a related protein cdk2, function late in the cycle while cdk4 and cdk6 are critically involved in G1 to S progression. CDK6 is essential for cell proliferation within the dentate gyrus of the hippocampus and the cuventricular zone of the lateral ventricles. Mutations in the gene results in microcephaly 12, primary, autosomal recessive (MCPH12).

Specifications

Antigen Phospho-CDK6 (Tyr24)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CDK6
Gene Accession No. Q00534
Gene Alias 5830411I20; AI504062; Cdk6; CDKN6; Cell division protein kinase 6; CR2 protein kinase; Crk2; cyclin dependent kinase 6; Cyclin-dependent kinase 6; MCPH12; MGC59692; PLSTIRE; Serine/threonine-protein kinase PLSTIRE
Gene Symbols CDK6
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human CDK6 around the phosphorylation site of Tyr24.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1021
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.