Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-CENTB1 (Ser554) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564529
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564529 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564529 Supplier Invitrogen™ Supplier No. PA564529
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-CENTB1 (Ser554) Polyclonal Antibody detects endogenous levels of CENTB1 only when phosphorylated at Ser554.

ACAP1, also known as Centaurin-beta-1 (CENTB1), is a 740 amino acid containing transport carrier belonging to the Centaurin family with three ANK repeats, an Arf-GAP domain, a BAR domain and a PH domain that binds to phospholipids. ACAP1, a GTPase-activating protein (GAP) for ADP ribosylation factor 6 (ARF6), is required for clathrin-dependent export of proteins from recycling endosomes to trans-Golgi network and cell surface. It mediates CENTB1-binding to phosphatidylinositol 3,5-bisphosphate (PIP2) or phosphatidylinositol 3,4,5-trisphosphate (PIP3) containing membranes. Along with ACAP2, ACAP1 is recruited to platelet-derived growth factor (PDGF) induced dorsal membrane ruffles in NIH 3T3 mouse fibroblasts, while over expression inhibited ruffle formation. It is widely expressed in most tissues with high expression in lung and spleen and low expression in heart, kidney, liver and pancreas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-CENTB1 (Ser554)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ACAP1
Gene Accession No. Q15027
Gene Alias Acap1; Arf GAP with coiled coil, ANK repeat and PH domains 1; arf-GAP with coiled-coil, ANK repeat and PH domain-containing protein 1; ArfGAP with coiled-coil, ankyrin repeat and PH domains 1; centaurin beta 1; centaurin, beta 1; centaurin-beta-1; Centb1; cnt-b1; KIAA0050
Gene Symbols ACAP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CENTB1 (Phospho-Ser554).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9744
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.