Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-CENTB1 (Ser554) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538753
Description
ACAP1 or Centaurin beta1 (CENTB1), a GTPase-activating protein, is a member of the ADP-ribosylation factor protein family defined by several common structural motifs including coiled coil, pleckstrin homology, Arf GAP, and three complete ankyrin-repeat domains. ACAP1 is widely expressed and functions as a Arf6GAP.
Specifications
| Phospho-CENTB1 (Ser554) | |
| Polyclonal | |
| Unconjugated | |
| ACAP1 | |
| ACAP1, centaurin beta 1, KIAA0050, Arf GAP with coiled coil, ANK repeat and PH domains 1, ArfGAP with coiled-coil, ankyrin repeat and PH domains 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9744 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q15027 | |
| ACAP1 | |
| A synthetic phosphopeptide derived from human Centaurin-beta 1 around the phosphorylation site of Ser554 (P-G-SP-L-R) | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction