Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-CEP55 (Ser425) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564661
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564661 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564661 Supplier Invitrogen™ Supplier No. PA564661
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-CEP55 (Ser425) Polyclonal Antibody detects endogenous levels of CEP55 only when phosphorylated at Ser425.

Plays a role in mitotic exit and cytokinesis (PubMed:16198290, PubMed:17853893). Recruits PDCD6IP and TSG101 to midbody during cytokinesis. Required for successful completion of cytokinesis (PubMed:17853893). Not required for microtubule nucleation (PubMed:16198290). Plays a role in the development of the brain and kidney (PubMed:28264986). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-CEP55 (Ser425)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CEP55
Gene Accession No. Q53EZ4
Gene Alias 1200008O12Rik; 2700032M20Rik; C10orf3; cancer/testis antigen 111; centrosomal protein 55; centrosomal protein 55kDa; centrosomal protein of 55 kDa; centrosomal protein of 55 kDa; LOW QUALITY PROTEIN: centrosomal protein of 55 kDa; centrosomal protein of 55 kDa-like protein; CEP55; CT111; RGD1305340; Up-regulated in colon cancer 6; URCC6
Gene Symbols CEP55
Host Species Rabbit
Immunogen A synthesized peptide derived from human CEP55 (Phospho-Ser425).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55165
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.