Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-CFTR (Ser737) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564505
Description
Phospho-CFTR (Ser737) Polyclonal Antibody detects endogenous levels of CFTR only when phosphorylated at Ser737.
Cystic Fibrosis (CF) is a common lethal genetic disease caused by mutations of the gene coding for the cystic fibrosis transmembrane conductance factor, a cAMP regulated chloride channel. Approximately 70% of all CF cases share the deletion of a phenylalanine at position 508 (delta F508) which results in abnormal chloride transport. Since the CF mutation is lethal, most often by lung and liver disease, it raises the question of why this genetic disease remains as common as it is. One possible explanation is that Salmonella typhi has been shown to use CFTR to enter intestinal epithelial cells and that delta F508 heterozygote and homozygote mice showed 86% and 100% reductions in S. typhi intestinal submucosal uptake.
Specifications
Phospho-CFTR (Ser737) | |
Polyclonal | |
Unconjugated | |
CFTR | |
ABC35; Abcc7; ATP Binding Cassette Superfamily C Member 7 (ABCC7); ATP-binding cassette sub-family C member 7; ATP-binding cassette transporter sub-family C member 7; ATP-binding cassette, subfamily c, member 7; AW495489; cAMP-dependent chloride channel; CF; CFTR; CFTR/MRP; Channel conductance-controlling ATPase; cystic fibrosis transmembrane conductance regulator; cystic fibrosis transmembrane conductance regulator (ATP-binding cassette sub-family C, member 7); cystic fibrosis transmembrane conductance regulator homolog; cystic fibrosis transmembrane conductance regulator homolog; ATP-binding cassette, subfamily c, member 7; dJ760C5.1; MRP7; RGD1561193; tcag7.78; TNR CFTR; TNR-CFTR | |
Rabbit | |
Affinity chromatography | |
RUO | |
1080, 12638, 24255 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P13569, P26361, P34158 | |
CFTR | |
A synthesized peptide derived from human CFTR (Phospho-Ser737). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction