Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-CFTR (Ser737) Polyclonal Antibody

Phospho-CFTR (Ser737) Polyclonal Antibody
Supplier: Thermo Scientific PA5105784
Description
Phospho-CFTR (Ser737) Polyclonal Antibody for Western Blot, IHC (P)
Specifications
| Phospho-CFTR (Ser737) | |
| Polyclonal | |
| Unconjugated | |
| CFTR | |
| ABC35; Abcc7; ATP Binding Cassette Superfamily C Member 7 (ABCC7); ATP-binding cassette sub-family C member 7; ATP-binding cassette transporter sub-family C member 7; ATP-binding cassette, subfamily c, member 7; AW495489; cAMP-dependent chloride channel; CF; CFTR; CFTR/MRP; Channel conductance-controlling ATPase; cystic fibrosis transmembrane conductance regulator; cystic fibrosis transmembrane conductance regulator (ATP-binding cassette sub-family C, member 7); cystic fibrosis transmembrane conductance regulator homolog; cystic fibrosis transmembrane conductance regulator homolog; ATP-binding cassette, subfamily c, member 7; dJ760C5.1; MRP7; RGD1561193; tcag7.78; TNR CFTR; TNR-CFTR | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 1080, 12638, 24255 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P13569, P26361, P34158 | |
| CFTR | |
| A synthesized peptide derived from human CFTR(Accession P13569), corresponding to amino acid residues around phosphorylated Ser737. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction