Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Cofilin (Tyr140) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986445
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6445 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6445 Supplier Bioss Supplier No. BS5258R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cofilin binds to F-actin and exhibits pH-sensitive F-actin depolymerizing activity. Regulates actin cytoskeleton dynamics. Important for normal progress through mitosis and normal cytokinesis. Plays a role in the regulation of cell morphology and cytoskeletal organization. Required for the up-regulation of atypical chemokine receptor ACKR2 from endosomal compartment to cell membrane, increasing its efficiency in chemokine uptake and degradation. Required for neural tube morphogenesis and neural crest cell migration.

Specifications

Antigen Phospho-Cofilin (Tyr140)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CFL1
Gene Accession No. P23528
Gene Alias 18 kDa phosphoprotein; AA959946; BOS_24828; CFL; Cfl1; CFL-1; Cof; COF1; cofilin; cofilin 1; cofilin 1 (non-muscle); cofilin 1, non-muscle; COFILIN protein; Cofilin, non-muscle isoform; Cofilin1; Cofilin-1; cofilin-1-like protein; epididymis secretory protein Li 15; HEL-S-15; n-cofilin; p18
Gene Symbols CFL1
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human Cofilin around the phosphorylation site of Tyr140.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1072
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.