Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Connexin 43/GJA1 (S368) Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Supplier: R&D Systems PPS046
This item is not returnable.
View return policy
Description
Prepared from rabbit serum by affinity purification via sequential chromatography on phospho and dephosphopeptide coupled columns.
Connexin 43/GJA1 Polyclonal specifically detects Connexin 43/GJA1 in Human, Mouse, Rat, Bovine, Canine, Chicken, Guinea Pig, Primate, Zebrafish samples. It is validated for Western Blot.Specifications
Connexin 43/GJA1 (p ser368) | |
Polyclonal | |
Western Blot 1:1000 dilution | |
GJA1 | |
Phosphopeptide corresponding to amino acid residues surrounding the phospho-S368 of Connexin 43/GJA1 | |
100 μL | |
Primary | |
This antibody is specific for the 43 kDa Connexin 43/GJA1 in Western blots of rat brain lysates. | |
For long-term storage, < -20degrees C is recommended. Product is stable at < -20degrees C for at least 1 year. |
Western Blot | |
Unconjugated | |
connexin 43, connexin-43, Cx43, CX43gap junction protein, alpha-like, DFNB38, Gap junction 43 kDa heart protein, gap junction alpha-1 protein, gap junction protein, alpha 1, 43kDa, gap junction protein, alpha 1, 43kDa (connexin 43), GJA1, GJAL, HSS, ODD, ODDD, ODOD, SDTY3 | |
Rabbit | |
Affinity Purified | |
RUO | |
2697 | |
Human, Mouse, Rat, Bovine, Canine, Chicken, Guinea Pig, Primate, Zebrafish | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction