Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-CREB/ATF1 (Ser133, Ser63) Monoclonal Antibody (10E9)
GREENER_CHOICE

Catalog No. PIMA1114
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA1114 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA1114 Supplier Invitrogen™ Supplier No. MA1114
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

This antibody recognizes CREB protein phosphorylated at serine 133 and ATF1 phosphorylated at serine 63. The antibody was purified from serum-free cell culture supernatant by subsequent thiophilic adsorption and size exclusion chromatography. Reconstitute with 1 mL H2O (15 min, RT) Aliquot and freeze in liquid nitrogen. Antibody can be stored frozen at -80°C for up to 1 year. Thaw aliquots at 37°C; thawed aliquots may be stored at 4°C for up to 3 months. Avoid repeated freeze/thaw cycles. This antibody was originally validated as part of a Thermo Scientific Cellomics High Content Screening Kit. The antibody sold separately may have slightly different performance and may need to be further optimized for the best results.

Cyclic AMP response element binding protein (CREB) is a basic/leucine zipper transcription factor that binds the cyclic AMP response element (CRE) and activates transcription in response to a variety of extracellular signals including neurotransmitters, hormones, membrane depolarization, and growth and neurotrophic factors. Activation of CREB is dependent upon the phosphorylation of serine 133. Phosphorylation occurs via p44/42 MAP kinase and p90RSK and also via p38 MAP kinase and MSK1. Although CREB will bind DNA independent of its phosphorylation state, only the phosphorylated form is competent as a transcription factor. CREB binding protein (CBP), a transcriptional coactivator that directly interacts with CREB, binds to CREB in the region of serine 133. CREB appears to play an important role in learning and memory.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-CREB/ATF1 (Ser133, Ser63)
Applications ChIP Assay, ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 10E9
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, PEG, sucrose and 0.09% sodium azide; pH 7.2
Gene ATF1
Gene Accession No. P16220, P18846, P81269, Q01147
Gene Alias 2310001E10Rik; 3526402H21Rik; Activating transcription factor 1; active transcription factor CREB; ATF1; AV083133; cAMP responsive element binding protein 1; cAMP-dependent transcription factor ATF-1; cAMP-response element-binding protein-1; cAMP-responsive element-binding protein 1; Creb; CREB1; Creb-1; cyclic adenosine 3',5'-monophosphate response element-binding protein CREB; cyclic AMP-dependent transcription factor ATF-1; Cyclic AMP-responsive element-binding protein 1; EWS-ATF1; FUS/ATF-1; Protein TREB36; Ser133; TCR-ATF1; transactivator protein; TREB36
Gene Symbols ATF1, CREB1
Host Species Mouse
Immunogen A synthetic phosphopeptide corresponding to amino acids 125-135 surrounding the Ser133 phosphorylation site of human CREB.
Purification Method Size-exclusion chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11908, 12912, 1385, 466, 486545, 607922
Target Species Canine, Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.