Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-Crk (Ser41) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA512738
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA512738 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA512738 Supplier Invitrogen™ Supplier No. PA512738
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with mouse and rat based on sequence homology.

The Crk-I and Crk-II forms differ in their biological activities. Crk-II has less transforming activity than Crk-I. Crk- II mediates attachment-induced MAPK8 activation, membrane ruffling and cell motility in a Rac-dependent manner. Involved in phagocytosis of apoptotic cells and cell motility via its interaction with DOCK1 and DOCK4. May regulate the EFNA5-EPHA3 signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Crk (Ser41)
Applications Dot Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Crk
Gene Accession No. P46108, Q16539
Gene Alias Adapter molecule crk; avian sarcoma virus CT10 (v-crk) oncogene homolog; c-Crk; CRK; CRK proto-oncogene, adaptor protein; CRK1; Crk3; Crk-I; CRKII; Crk-II; CrkIII; Crk-III; Crko; CSAID-binding protein; Csaids binding protein; CSBP; Csbp1; CSBP2; CSPB1; CT-10 related kinase 3; cytokine suppressive anti-inflammatory drug binding protein; cytokine suppressive anti-inflammatory drug binding protein 1; cytokine suppressive anti-inflammatory drug-binding protein; cytokine-supressive anti-inflammatory drug binding protein; Exip; Hog; I79_016605; MAP kinase 14; MAP kinase 2; MAP kinase Mxi2; MAP kinase p38 alpha; MAPK 14; MAPK14; MAX-interacting protein 2; mitogen activated protein kinase 14; mitogen-activated protein kinase 14; mitogen-activated protein kinase 14A; mitogen-activated protein kinase p38 alpha; MXI2; p38; p38 alpha; p38 MAP kinase; p38 MAP kinase alpha; p38 MAPK; p38 mitogen activated protein kinase; p38a; p38ALPHA; p38-alpha; p38alpha Exip; p38Hog; p38MAPK; PRKM14; PRKM15; Proto-oncogene c-Crk; reactive kinase; RK; SAPK2A; stress-activated protein kinase 2A; tRNA synthetase cofactor p38; v-crk avian sarcoma virus CT10 oncogene homolog; v-crk sarcoma virus CT10 oncogene homolog; v-crk sarcoma virus CT10 oncogene homolog (avian); v-crk sarcoma virus CT10 oncogene-like protein
Gene Symbols Crk, Mapk14
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S41 of human CRK.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1398, 1432
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.