Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-CRK (Ser41) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986448
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6448 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6448 Supplier Bioss Supplier No. BS5264R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The Crk-I and Crk-II forms differ in their biological activities. Crk-II has less transforming activity than Crk-I. Crk- II mediates attachment-induced MAPK8 activation, membrane ruffling and cell motility in a Rac-dependent manner. Involved in phagocytosis of apoptotic cells and cell motility via its interaction with DOCK1 and DOCK4. May regulate the EFNA5-EPHA3 signaling.

Specifications

Antigen Phospho-CRK (Ser41)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Crk
Gene Accession No. P46108
Gene Alias Adapter molecule crk; avian sarcoma virus CT10 (v-crk) oncogene homolog; c-Crk; Crk; CRK proto-oncogene, adaptor protein; Crk3; Crk-I; CRKII; Crk-II; CrkIII; Crk-III; Crko; CT-10 related kinase 3; p38; Proto-oncogene c-Crk; v-crk avian sarcoma virus CT10 oncogene homolog; v-crk sarcoma virus CT10 oncogene homolog; v-crk sarcoma virus CT10 oncogene homolog (avian); v-crk sarcoma virus CT10 oncogene-like protein
Gene Symbols Crk
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human CRK around the phosphorylation site of Ser41 [DS (p-S)TS].
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1398
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.