Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-CRMP2 (Ser522) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143675
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143675 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143675 Supplier Invitrogen™ Supplier No. PA5143675
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects a 70 kDa* protein corresponding to the molecular mass of CRMP2 on SDS-PAGE immunoblots of human mouse brain, and this band is removed after lambda phosphatase treatment. In addition, this reactivity can be specifically blocked using phospho-CRMP2 (Ser-522) peptide (CX2195).

Necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. Plays a role in axon guidance, neuronal growth cone collapse and cell migration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-CRMP2 (Ser522)
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene DPYSL2
Gene Accession No. O08553, P47942, Q16555
Gene Alias AI851130; collapsin response mediator protein 2; collapsin response mediator protein hCRMP-2; CRMP2; CRMP-2; DHPRP2; dihydropyrimidinase like 2; dihydropyrimidinase-like 2; dihydropyrimidinase-related protein 2; DPYL2; Dpysl2; DRP2; DRP-2; Musunc33; N2A3; Neural-specific protein NSP60; TOAD-64; turned on after division 64; turned on after division 64 kDa protein; turned on after division, 64 kDa protein; ULIP2; ULIP-2; unc-33-like phosphoprotein 2; Unknown (protein for MGC:137406)
Gene Symbols DPYSL2
Host Species Rabbit
Immunogen Phospho-CRMP2 (Ser-522) synthetic peptide (coupled to carrier protein) corresponding to amino acids surrounding Ser-522 in human CRMP2. This sequence is conserved in rat and mouse CRMP2, and has homology to the conserved site in CRMP1 and CRMP4, but is not conserved in CRMP3 and CRMP5.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12934, 1808, 25416
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.