Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-CRMP2 (Ser522) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537550
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537550 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537550 Supplier Invitrogen™ Supplier No. PA537550
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control for Western blot is HUVEC cells.

Necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. Plays a role in axon guidance, neuronal growth cone collapse and cell migration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-CRMP2 (Ser522)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DPYSL2
Gene Accession No. Q16555
Gene Alias AI851130; collapsin response mediator protein 2; collapsin response mediator protein hCRMP-2; CRMP2; CRMP-2; DHPRP2; dihydropyrimidinase like 2; dihydropyrimidinase-like 2; dihydropyrimidinase-related protein 2; DPYL2; Dpysl2; DRP2; DRP-2; Musunc33; N2A3; Neural-specific protein NSP60; TOAD-64; turned on after division 64; turned on after division 64 kDa protein; turned on after division, 64 kDa protein; ULIP2; ULIP-2; unc-33-like phosphoprotein 2; Unknown (protein for MGC:137406)
Gene Symbols DPYSL2
Host Species Rabbit
Immunogen Peptide sequence around phosphorylation site of Serine 522(K-T-S(p)-P-A) derived from Human CRMP-2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1808
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.