Learn More
Invitrogen™ Phospho-CRMP2 (Thr555) Monoclonal Antibody (M539)

Description
The antibody detects a 70 kDa* protein corresponding to CRMP2 on immunoblots of rat PC12 or mouse C2C12 cells treated with calyculin A. This reactivity is not observed after lambda phosphatase dephosphorylation of Thr-555.
Specifications
Specifications
| Antigen | Phospho-CRMP2 (Thr555) |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | M539 |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide |
| Gene | DPYSL2 |
| Gene Accession No. | O08553, P47942, Q16555 |
| Gene Alias | AI851130; collapsin response mediator protein 2; collapsin response mediator protein hCRMP-2; CRMP2; CRMP-2; DHPRP2; dihydropyrimidinase like 2; dihydropyrimidinase-like 2; dihydropyrimidinase-related protein 2; DPYL2; Dpysl2; DRP2; DRP-2; Musunc33; N2A3; Neural-specific protein NSP60; TOAD-64; turned on after division 64; turned on after division 64 kDa protein; turned on after division, 64 kDa protein; ULIP2; ULIP-2; unc-33-like phosphoprotein 2; Unknown (protein for MGC:137406) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.