Learn More
Invitrogen™ Phospho-CRMP2 (Thr555) Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5143676
Description
This antibody was cross-adsorbed to unphosphorylated CRMP2 (Thr-555) before affinity purification using phospho-CRMP2 (Thr-555) peptide (without carrier). The antibody detects a 70 kDa* protein corresponding to CRMP2 on immunoblots of rat PC12 or mouse C2C12 cells treated with calyculin A. This reactivity can be specifically blocked using phospho-CRMP2 (Thr-555) peptide (CX2255).
Necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. Plays a role in axon guidance, neuronal growth cone collapse and cell migration.
Specifications
Phospho-CRMP2 (Thr555) | |
Polyclonal | |
Unconjugated | |
DPYSL2 | |
AI851130; collapsin response mediator protein 2; collapsin response mediator protein hCRMP-2; CRMP2; CRMP-2; DHPRP2; dihydropyrimidinase like 2; dihydropyrimidinase-like 2; dihydropyrimidinase-related protein 2; DPYL2; Dpysl2; DRP2; DRP-2; Musunc33; N2A3; Neural-specific protein NSP60; TOAD-64; turned on after division 64; turned on after division 64 kDa protein; turned on after division, 64 kDa protein; ULIP2; ULIP-2; unc-33-like phosphoprotein 2; Unknown (protein for MGC:137406) | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12934, 1808, 25416 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot, Immunocytochemistry | |
~0.4 mg/mL | |
PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
O08553, P47942, Q16555 | |
DPYSL2 | |
Phospho-CRMP2 (Thr-555) synthetic peptide (coupled to carrier protein) corresponding to amino acids surrounding Thr-555 in human CRMP2. This sequence is conserved in rat and mouse CRMP2 and the phospho-site is not conserved in other CRMP family members. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.