Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-CSF1R (Tyr723) Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS3077R

Description
CSF1R (colony stimulating factor 1 receptor) is a cytokine that controls the production, differentiation, and function of macrophages. CSF1R mediates most if not all of the biological effects of CSF1. Ligand binding activates the receptor kinase through a process of oligomerization and transphosphorylation. CSF1R is a tyrosine kinase transmembrane receptor and member of the CSF1/PDGF receptor family of tyrosine-protein kinases. Mutations in the gene encoding CSF1R have been associated with a predisposition to myeloid malignancy.Tyrosine-protein kinase that acts as cell-surface receptor for CSF1 and IL34 and plays an essential role in the regulation of survival, proliferation and differentiation of hematopoietic precursor cells, especially mononuclear phagocytes, such as macrophages and monocytes. Promotes the release of proinflammatory chemokines in response to IL34 and CSF1, and plays an important role in innate immunity and in inflammatory processes. The first intron of the CSF1R gene contains a transcriptionally inactive ribosomal protein L7 processed pseudogene oriented in the opposite direction. Mutations in the CSF1R gene have been associated with a predisposition to myeloid malignancy.Specifications
Phospho-CSF1R (Tyr723) | |
Polyclonal | |
Unconjugated | |
Csf1r | |
AI323359; CD115; CD115 antigen; C-FMS; colony stimulating factor 1 receptor; CSF-1 receptor; CSF1R; CSF-1R; CSF-1-R; Csfmr; CSFR; EC 2.7.10.1; FIM2; Fim-2; Fms; fms proto-oncogene; HDLS; kinase CSFR; macrophage colony stimulating factor I receptor; Macrophage colony-stimulating factor 1 receptor; McDonough feline sarcoma viral (v-fms) oncogene homolog; M-CSFR; M-CSF-R; mrfms; OTTHUMP00000224093; PDGFRB; Proto-oncogene c-Fms; proto-oncogene fms | |
Rabbit | |
Protein A | |
RUO | |
1436 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P07333 | |
Csf1r | |
KLH conjugated synthetic phosphopeptide derived from human CSF1R around the phosphorylation site of tyrosine 723. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction