Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-CtBP1/CtBP2 (Ser158, Ser164) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564665
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564665 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564665 Supplier Invitrogen™ Supplier No. PA564665
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-CtBP1/CtBP2 (Ser158, 164) Polyclonal Antibody detects endogenous levels of CtBP1/CtBP2 only when phosphorylated at Ser158, 164.

This gene encodes a protein that binds to the C-terminus of adenovirus E1A proteins. This phosphoprotein is a transcriptional repressor and may play a role during cellular proliferation. This protein and the product of a second closely related gene, CTBP2, can dimerize. Both proteins can also interact with a polycomb group protein complex which participates in regulation of gene expression during development. Alternative splicing of transcripts from this gene results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-CtBP1/CtBP2 (Ser158, Ser164)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CTBP2
Gene Accession No. P56545, Q13363
Gene Alias 50 kDa BFA-dependent ADP-ribosylation substrate; 50-kDa BFA-induced ADP-ribosylated substrate; 50-kDaBFA-inducedADP-ribosylatedsubstrate; AA407280; Bars; BARS-50; brefeldin A-ADP-riboslyated substrate; brefeldin A-ribosylated substrate; CTBP; Ctbp1; CtBP1-L; CtBP1-S; CTBP2; Ctbp3; CtBP3/BARS; C-terminal binding protein 1; C-terminal binding protein 2; C-terminal-binding protein 1; C-terminal-binding protein 2; C-terminal-binding protein 3; C-terminus binding protein 3/brefeldin A (BFA) adenosine diphosphate-ribosylated substrate; D4S115h; D5H4S115; D5H4S115E; D7Ertd45e; gene trap ROSA b-geo 6; Gtrgeo6; ribeye
Gene Symbols CTBP1, CTBP2
Host Species Rabbit
Immunogen A synthesized peptide derived from human CtBP1/2 (Phospho-Ser158/164).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1487, 1488
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.