Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-CtIP (Ser664) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105832
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105832 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105832 Supplier Invitrogen™ Supplier No. PA5105832
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of CtIP only when phosphorylated at Ser664.

CtIP, a 125 kD protein, was originally found interacting with a transcription repressor, CtBP, through the PLDLS motif (CtBP-Interacting Protein) thus suggested a role in transcription. Studies have shown that CtIP also interacts with BRCA1 protein through the c-terminus BRCT domains also suggested that CtIP is a potential tumor suppressor. This CtIP-BRCA1 interaction can be disrupted by DNA damaging agents including UV or gamma-irradiation. Li et al (Nature 406, 210 - 215 (2000)) have shown that ATM phosphorylates CtIP at serine residues 664 and 745, and mutation of these sites abrogates the dissociation of BRCA1 from CtIP, resulting in persistent repression of BRCA1-dependent induction of GADD45 upon ionizing radiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-CtIP (Ser664)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Rbbp8
Gene Accession No. B1WC58, Q80YR6, Q99708
Gene Alias 9930104E21Rik; COM1; CtBP-interacting protein; CtIP; DNA endonuclease RBBP8; JWDS; RB binding protein 8, endonuclease; RBBP8; RBBP-8; retinoblastoma binding protein 8; retinoblastoma-binding protein 8; Retinoblastoma-interacting protein and myosin-like; RGD1308872; RIM; SAE2; SCKL2; Sporulation in the absence of SPO11 protein 2 homolog
Gene Symbols Rbbp8
Host Species Rabbit
Immunogen A synthesized peptide derived from human RBBP8(Accession Q99708), corresponding to amino acid residues around phosphorylated Ser664.
Purification Method sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225182, 291787, 5932
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.