Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-CTNNAL1 (Tyr436) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105367
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105367 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105367 Supplier Invitrogen™ Supplier No. PA5105367
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of CTNNAL1 only when phosphorylated at Tyr436.

May modulate the Rho pathway signaling by providing a scaffold for the Lbc Rho guanine nucleotide exchange factor (ARHGEF1).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-CTNNAL1 (Tyr436)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CTNNAL1
Gene Accession No. O88327, Q9UBT7
Gene Alias ACRP; AI616177; alpha2-catulin; alpha-catenin related protein; alpha-catenin-related protein; alpha-CATU; Alpha-catulin; AW545119; C86009; catenin (cadherin associated protein), alpha-like 1; catenin (cadherin-associated protein), alpha-like 1; catenin alpha like 1; catenin alpha-like 1; catenin alpha-like protein 1; Catnal1; CLLP; CTNNAL1; RP11-115J22.1
Gene Symbols CTNNAL1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CTNNAL1(Accession Q9UBT7), corresponding to amino acid residues around phosphorylated Tyr436.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 298019, 54366, 8727
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.