Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-CUX1 (Ser1237) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564669
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564669 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564669 Supplier Invitrogen™ Supplier No. PA564669
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-CUX1 (Ser1237) Polyclonal Antibody detects endogenous levels of CUX1 only when phosphorylated at Ser1237.

The protein encoded by this gene is a member of the homeodomain family of DNA binding proteins. It may regulate gene expression, morphogenesis, and differentiation and it may also play a role in the cell cycle progession. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-CUX1 (Ser1237)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CUX1
Gene Accession No. P39880, P53565, P70403, Q13948
Gene Alias CASP; CCAAT displacement protein; CDP; CDP/Cut; CDP1; CDP2; Cdpl1; Clox; COY1; cut homolog; cut like homeobox 1; CUTL1; cut-like 1; cut-like homeobox 1; Cux; Cux/CDP; Cux/CDP homeoprotein; CUX1; Cux-1; golgi integral membrane protein 6; GOLIM6; Homeobox protein cut-like 1; homeobox protein cux-1; Kiaa4047; Nbla10317; p100; p110; p200; p75; Protein CASP; putative protein product of Nbla10317
Gene Symbols CUX1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CUTL1 (Phospho-Ser1237).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116639, 13047, 1523
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.