Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-CYLD (Ser418) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105234
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105234 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105234 Supplier Invitrogen™ Supplier No. PA5105234
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of CYLD only when phosphorylated at Ser418.

CYLD is a 956 aa, cytoplasmic, deubiquitinating enzyme belonging to the ubiquitin carboxy-terminal hydrolases (UCH) family of proteins with three cytoskeletal-associated protein-glycine-conserved (CAP-GLY) domains, a proline rich region, a SH3 binding domain and a sequence homology to catalytic domain of UCH. CYLD is identified as a tumor suppressor protein affecting the JNK signaling pathway. CYLD is a negative regulator of TRAF2 and NF-kappa-B signaling pathway and is also known to have receptor-dependent role in regulating the I-kappa-B kinase pathway. It also has a deubiquitinating activity that is directed towards non-Lys-48-linked polyubiquitin chains and TRAP1 is a novel substrate for deubiquitination. Mutated CYLD is known to be associated with cylindromatosis, multiple familial trichoepithelioma, and Brooke-Spiegler syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-CYLD (Ser418)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CYLD
Gene Accession No. Q66H62, Q9NQC7
Gene Alias 2010013M14Rik; 2900009M21Rik; BRSS; C130039D01Rik; CDMT; Cyld; CYLD lysine 63 deubiquitinase; CYLD1; CYLDI; cylindromatosis (turban tumor syndrome); deubiquitinating enzyme CYLD; EAC; FLJ20180; FLJ31664; FLJ78684; HSPC057; KIAA0849; LRRGT00003; MFT; MFT1; mKIAA0849; probable ubiquitin carboxyl-terminal hydrolase CYLD; retinitis pigmentosa 1 homolog; Rp1; Rp1h; SBS; TEM; ubiquitin carboxyl-terminal hydrolase CYLD; ubiquitin specific peptidase like 2; ubiquitin thioesterase CYLD; ubiquitin thiolesterase CYLD; ubiquitin-specific-processing protease CYLD; Unknown (protein for MGC:137952); USPL2
Gene Symbols CYLD
Host Species Rabbit
Immunogen A synthesized peptide derived from human CYLD(Accession Q9NQC7), corresponding to amino acid residues around phosphorylated Ser418.
Purification Method Sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1540, 312937
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.