Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
phospho-cytokeratin-18 (K18) (Ser33), Mouse anti-Human, Clone: IB4, Millipore Sigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABT875100UG
Description
Anti-phospho-cytokeratin-18 (K18) (Ser33), clone IB4, Cat. No. MABT875, is a mouse monoclonal antibody that detects type I cytoskeletal-18 and has been tested for use in Immunocytochemistry, Immunofluorescence, Immunoprecipitation, and Western Blotting.
Keratin, type I cytoskeletal 18 (UniProt: P05783; also known as Cell proliferation-inducing gene 46 protein, Cytokeratin-18, CK-18, Keratin-18, K18) is encoded by the KRT18 (also known as CYK18, PIG46) gene (Gene ID: 3875) in human. Cytokeratin-18 is a type I, acidic, protein that is expressed in colon, placenta, and liver. Higher expression levels have been reported in lymph nodes of breast carcinoma. Cytokeratin-18 is involved in the uptake of thrombin-antithrombin complexes by hepatic cells. Upon phosphorylation, it plays a role in filament reorganization. It is proteolytically cleaved by caspases during epithelial cell apoptosis and this cleavage is shown to occur at Asp-238. Phosphorylation of cytokeratin-18 at serine 33 is shown to increase during mitosis in cultured cells and in regenerating liver. Serine 33 phosphorylation is considered to be essential for its association with 14-3-3 proteins and has a role in keratin organization and distribution. Cytokeratin-18 can undergo O-glycosylation, which results in its increased solubility and reduction in its stability. Mutations in KRT18 gene are known to cause liver cirrhosis characterized by severe panlobular liver-cell swelling with Mallory body formation, prominent pericellular fibrosis, and marked deposits of copper.
Specifications
| phospho-cytokeratin-18 (K18) (Ser33) | |
| Monoclonal | |
| Unconjugated | |
| Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
| KRT18;CYK18;PIG46 | |
| KLH-conjugated linear peptide corresponding to 13 amino acids surrounding phosphorylated serine 33 (initiator methionine removed) from the N-terminal region of human cytokeratin-18. | |
| 100 μg | |
| Cell Structure | |
| NP_000215 | |
| Human | |
| Purified |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| IB4 | |
| Immunocytochemistry Analysis: A representative lot detected phospho-cytokeratin-18 (K18) (Ser33) in immunocytochemistry applications (Ku, N.O., et. al. (1998). EMBO J. 17(7):1892-906).Immunoprecipitation Analysis: A representative lot immunoprecipitated phospho-cytokeratin-18 (K18) (Ser33) in immunoprecipitation applications (Ku, N.O., et. al. (1998). EMBO J. 17(7):1892-906).Immunofluorescence Analysis: A representative lot detected phospho-cytokeratin-18 (K18) (Ser33) in Immunofluorescence applications (Ku, N.O., et. al. (1998). EMBO J. 17(7):1892-906).Western Blotting Analysis: A representative lot detected phospho-cytokeratin-18 (K18) (Ser33) in Western Blotting applications (Yoon, K.H., et. al. (2001). J Cell Biol. 153(3):503-16). | |
| Keratin type I cytoskeletal 18;Cell proliferation-inducing gene 46 protein;CK-18;Keratin-18;K18 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Clone IB4 specifically detects human cytokeratin-18 phosphorylated on serine 33. | |
| Stable for 1 year at 2-8°C from date of receipt. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction